Assay Details
| Gene Symbol: ITGB1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319066.1 | NP_001305995.1 | 3 - 4 | 491 | 104 | |
| NM_001319067.1 | NP_001305996.1 | 3 - 4 | 393 | 104 | ||
| NM_001319068.1 | NP_001305997.1 | 3 - 4 | 364 | 104 | ||
| NM_004763.4 | NP_004754.1 | 3 - 4 | 379 | 104 | ||
| NM_022334.4 | NP_071729.1 | 3 - 4 | 379 | 104 | ||
| NR_134951.1 | 3 - 4 | 368 | 104 | |||
| NR_134952.1 | 3 - 4 | 379 | 104 | |||
| NR_134953.1 | 2 - 3 | 272 | 104 | |||
| NR_134954.1 | 3 - 4 | 379 | 104 | |||
| NR_134955.1 | 3 - 4 | 379 | 104 | |||
| NR_134957.1 | 2 - 3 | 272 | 104 | |||
| XM_005246183.4 | XP_005246240.1 | 4 - 5 | 1065 | 104 | ||
| XM_005246184.4 | XP_005246241.1 | 4 - 5 | 504 | 104 | ||
| XM_005246185.4 | XP_005246242.1 | 4 - 5 | 377 | 104 | ||
| XM_005246189.4 | XP_005246246.1 | 4 - 5 | 1030 | 104 | ||
| XM_006711903.3 | XP_006711966.1 | 3 - 4 | 609 | 104 | ||
| XM_011510416.2 | XP_011508718.1 | 4 - 5 | 699 | 104 | ||
| XM_017005267.1 | XP_016860756.1 | 4 - 5 | 2194 | 104 | ||
| XM_017005268.1 | XP_016860757.1 | 4 - 5 | 369 | 104 | ||
| XM_017005269.1 | XP_016860758.1 | 3 - 4 | 920 | 104 | ||
| XM_017005270.1 | XP_016860759.1 | 4 - 5 | 2203 | 104 | ||
| GenBank mRNA | AF012023.1 | - | 3 - 4 | 315 | 104 | |
| AF012024.1 | - | 3 - 4 | 315 | 104 | ||
| AI144519.1 | - | 3 - 4 | 405 | 104 | ||
| AI336197.1 | - | 3 - 4 | 307 | 104 | ||
| AK092115.1 | - | 266 | 104 | |||
| AK093179.1 | - | 3 - 4 | 953 | 104 | ||
| AK289817.1 | - | 3 - 4 | 292 | 104 | ||
| BC012264.1 | - | 3 - 4 | 271 | 3863540 | 104 | |
| BG032225.1 | - | 3 - 4 | 272 | 104 | ||
| BM823938.1 | - | 2 - 3 | 99 | 104 | ||
| BQ421551.1 | - | 3 - 4 | 263 | 104 | ||
| BQ425555.1 | - | 3 - 4 | 324 | 104 | ||
| BU520903.1 | - | 3 - 4 | 338 | 104 | ||
| CR457006.1 | - | 2 - 3 | 147 | 104 | ||
| CT002942.1 | - | 2 - 3 | 180 | 104 | ||
| DA087859.1 | - | 3 - 4 | 267 | 104 | ||
| DA751320.1 | - | 2 - 3 | 156 | 104 | ||
| DB167886.1 | - | 3 - 4 | 274 | 104 | ||
More Information
Set Membership: |
|

ITGB1BP1
Genomic Map