Assay Details
| Gene Symbol: PRMT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011523121.2 | XP_011521423.1 | 2 - 3 | 214 | 74 | |
| XM_011523124.2 | XP_011521426.1 | 3 - 4 | 393 | 74 | ||
| XM_011523125.2 | XP_011521427.1 | 2 - 3 | 235 | 74 | ||
| XM_011523126.2 | XP_011521428.1 | 2 - 3 | 239 | 74 | ||
| XM_011523131.2 | XP_011521433.1 | 2 - 3 | 214 | 74 | ||
| XM_017023302.1 | XP_016878791.1 | 2 - 3 | 235 | 74 | ||
| XM_017023303.1 | XP_016878792.1 | 3 - 4 | 393 | 74 | ||
| XM_017023305.1 | XP_016878794.1 | 2 - 3 | 214 | 74 | ||
| XM_017023307.1 | XP_016878796.1 | 3 - 4 | 393 | 74 | ||
| XM_017023308.1 | XP_016878797.1 | 3 - 4 | 407 | 74 | ||
| XM_017023309.1 | XP_016878798.1 | 2 - 3 | 235 | 74 | ||
| XM_017023310.1 | XP_016878799.1 | 2 - 3 | 259 | 74 | ||
| XM_017023313.1 | XP_016878802.1 | 3 - 4 | 382 | 74 | ||
| XM_017023315.1 | XP_016878804.1 | 3 - 4 | 382 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011523121.2, XM_011523124.2, XM_011523125.2, XM_011523126.2, XM_011523131.2, XM_017023302.1, XM_017023303.1, XM_017023305.1, XM_017023307.1, XM_017023308.1, XM_017023309.1, XM_017023310.1, XM_017023313.1, XM_017023315.1 transcripts.
Set Membership: |
|

PRMT7
Genomic Map