Assay Details
| Gene Symbol: PRMT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017023300.1 | XP_016878789.1 | 11 - 12 | 1527 | 83 | |
| XM_017023301.1 | XP_016878790.1 | 11 - 12 | 1527 | 83 | ||
| XM_017023306.1 | XP_016878795.1 | 11 - 12 | 1527 | 83 | ||
| XM_017023312.1 | XP_016878801.1 | 10 - 11 | 1400 | 83 | ||
| GenBank mRNA | AB067520.1 | - | 5 - 6 | 1537 | 83 | |
More Information
Set Membership: |
|

PRMT7
Genomic Map