Assay Details
| Gene Symbol: LRRC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019594.3 | NP_062540.2 | 1 - 2 | 144 | 147 | |
| GenBank mRNA | DA243590.1 | - | 1 - 2 | 144 | 147 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000704.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_019594.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09008693.hg.1 | TC09000704.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008693.hg.1 | ENST00000372599.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008693.hg.1 | ENST00000372600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008693.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008693.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000483638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000483638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000492784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000492784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | ENST00000492784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008691.hg.1 | TC09000704.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008691.hg.1 | ENST00000372600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008691.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008691.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008696.hg.1 | TC09000704.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008696.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008696.hg.1 | ENST00000372600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008696.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008696.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159948.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159948.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159948.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159948.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159948.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159948.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159948.hg.1 | TC0900008891.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900159950.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159950.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159950.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159950.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159950.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159950.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159950.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159950.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159950.hg.1 | TC0900008891.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372599.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000483638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000483638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000492784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000492784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | ENST00000492784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_001127245.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008891.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | ENST00000483638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | ENST00000259324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | ENST00000372600.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | ENST00000483638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | NM_001127244.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | NM_019594.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159953.hg.1 | TC0900008891.hg.1 | Clariom D TC |

LRRC8A
Genomic Map