Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003014.hg.1 |
ENST00000332123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003014.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003014.hg.1 |
TC10000262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003017.hg.1 |
ENST00000332123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003017.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003017.hg.1 |
TC10000262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003013.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003013.hg.1 |
TC10000262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003015.hg.1 |
ENST00000332123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003015.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003015.hg.1 |
TC10000262.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000150180.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150180.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150180.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150180.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150180.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150180.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150180.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150180.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150180.hg.1 |
TC1000007382.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000150181.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150181.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150181.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150181.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150181.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150181.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150181.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150181.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150181.hg.1 |
TC1000007382.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007382.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150187.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150187.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150187.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150187.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150187.hg.1 |
ENST00000424751.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150187.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150187.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150187.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150187.hg.1 |
TC1000007382.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000150178.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150178.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150178.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150178.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150178.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150178.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150178.hg.1 |
TC1000007382.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000150176.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150176.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150176.hg.1 |
TC1000007382.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000150177.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150177.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150177.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150177.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150177.hg.1 |
ENST00000429940.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150177.hg.1 |
NR_026827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150177.hg.1 |
TC1000007382.hg.1 |
Clariom D TC |