Assay Details
| Gene Symbol: LINC00839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC007394.2 | - | 3 - 4 | 679 | 93 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000424751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | ENST00000429940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007382.hg.1 | NR_026827.1 | Refseq |

LINC00839
Genomic Map