Assay Details
| Gene Symbol: EGFL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167890.1 | NP_001161362.1 | 8 - 9 | 1392 | 80 | |
| NM_015507.3 | NP_056322.2 | 8 - 9 | 1392 | 80 | ||
| GenBank mRNA | AF186084.1 | - | 8 - 9 | 1250 | 80 | |
| AF193055.1 | - | 8 - 9 | 1317 | 80 | ||
| AJ245671.1 | - | 8 - 9 | 1345 | 80 | ||
| AK075214.1 | - | 8 - 9 | 1362 | 80 | ||
| AK300236.1 | - | 7 - 8 | 1020 | 80 | ||
| AK315018.1 | - | 8 - 9 | 1230 | 80 | ||
| AL117610.1 | - | 8 - 9 | 1122 | 80 | ||
| AY358333.1 | - | 8 - 9 | 1238 | 80 | ||
| BC038587.1 | - | 8 - 9 | 1201 | 5767603 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000057.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000473826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000473826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | ENST00000473826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000772.hg.1 | TC0X000057.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000772.hg.1 | ENST00000361306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000772.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000772.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000772.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000473826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000473826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | ENST00000473826.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006651.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145486.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145486.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145486.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145486.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145486.hg.1 | ENST00000361306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145486.hg.1 | ENST00000380602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145486.hg.1 | NM_001167890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145486.hg.1 | NM_015507.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145486.hg.1 | TC0X00006651.hg.1 | Clariom D TC |

EGFL6
Genomic Map