Assay Details
| Gene Symbol: EGFL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167890.1 | NP_001161362.1 | 9 - 10 | 1467 | 71 | |
| GenBank mRNA | AJ245671.1 | - | 9 - 10 | 1420 | 71 | |
| AK300236.1 | - | 8 - 9 | 1095 | 71 | ||
More Information
Set Membership: |
|

EGFL6
Genomic Map