Assay Details
| Gene Symbol: TSPYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022117.3 | NP_071400.1 | 2 - 3 | 1025 | 74 | |
| XM_006724592.3 | XP_006724655.1 | 2 - 3 | 1058 | 74 | ||
| GenBank mRNA | AB015345.1 | - | 2 - 3 | 265 | 74 | |
| AF254794.1 | - | 2 - 3 | 764 | 74 | ||
| AF273046.1 | - | 2 - 3 | 1015 | 74 | ||
| AK292462.1 | - | 2 - 3 | 1018 | 74 | ||
| AL713652.1 | - | 2 - 3 | 1004 | 74 | ||
| AY040871.1 | - | 2 - 3 | 1014 | 74 | ||
| BC024270.1 | - | 2 - 3 | 1005 | 2819684 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000310.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
negative regulation of DNA replication
regulation of signal transduction
covalent chromatin modification
negative regulation of cell growth
IRE1-mediated unfolded protein response
negative regulation of cell cycle
regulation of protein kinase activity
rDNA binding
nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
negative regulation of DNA replication
regulation of signal transduction
covalent chromatin modification
negative regulation of cell growth
IRE1-mediated unfolded protein response
negative regulation of cell cycle
regulation of protein kinase activity
rDNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004153.hg.1 | TC0X000310.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004153.hg.1 | ENST00000375442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004153.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150320.hg.1 | TC0X00007310.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000375442.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000463525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000553557.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000556808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000578306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | ENST00000579390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007310.hg.1 | NM_022117.3 | Refseq |

TSPYL2
Genomic Map