Assay Details
| Gene Symbol: SERAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032861.3 | NP_116250.3 | 13 - 14 | 1549 | 85 | |
| XM_006715586.2 | XP_006715649.1 | 12 - 13 | 1478 | 85 | ||
| XM_011536196.2 | XP_011534498.1 | 12 - 13 | 1572 | 85 | ||
| XM_011536198.2 | XP_011534500.1 | 11 - 12 | 1474 | 85 | ||
| GenBank mRNA | AK027823.1 | - | 13 - 14 | 1539 | 85 | |
| AK125878.1 | - | 14 - 15 | 2741 | 85 | ||
| BC001705.1 | - | 13 - 14 | 1506 | 3350731 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

SERAC1
Genomic Map