Assay Details
| Gene Symbol: PRPF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003675.3 | NP_003666.1 | 7 - 8 | 868 | 116 | |
| XM_011519754.2 | XP_011518056.1 | 8 - 9 | 993 | 116 | ||
| GenBank mRNA | AK314814.1 | - | 7 - 8 | 843 | 116 | |
| BC000794.1 | - | 7 - 8 | 790 | 3451639 | 116 | |
| U51990.1 | - | 7 - 8 | 792 | 116 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000102.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001152.hg.1 | ENST00000378544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001152.hg.1 | ENST00000378572.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001152.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001152.hg.1 | TC10000102.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000320054.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012429.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185095.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185095.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185095.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185095.hg.1 | ENST00000378572.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185095.hg.1 | ENST00000417658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185095.hg.1 | NM_003675.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185095.hg.1 | TC1000012429.hg.1 | Clariom D TC |

PRPF18
Genomic Map