Assay Details
| Gene Symbol: TDRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146071.1 | NP_001139543.1 | 2 - 3 | 636 | 122 | |
| NM_030794.2 | NP_110421.1 | 2 - 3 | 636 | 122 | ||
| XM_005266560.2 | XP_005266617.1 | 2 - 3 | 594 | 122 | ||
| GenBank mRNA | AK024660.1 | - | 2 - 3 | 105 | 122 | |
| BC030514.1 | - | 2 - 3 | 158 | 4998330 | 122 | |
| BX537910.1 | - | 2 - 3 | 636 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK024660.1, BC030514.1, BX537910.1, NM_001146071.1, NM_030794.2, XM_005266560.2 transcripts.
Set Membership: |
|

TDRD3
Genomic Map