Assay Details
| Gene Symbol: PIGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004278.3 | NP_004269.1 | 5 - 6 | 562 | 69 | |
| XM_017025352.1 | XP_016880841.1 | 5 - 6 | 548 | 69 | ||
| XM_017025353.1 | XP_016880842.1 | 5 - 6 | 548 | 69 | ||
| GenBank mRNA | AB017165.1 | - | 5 - 6 | 535 | 69 | |
| AK292932.1 | - | 5 - 6 | 547 | 69 | ||
| BC068197.1 | - | 5 - 6 | 541 | 6711166 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000184.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000463810.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000463810.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000463810.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000470116.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000470116.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000470116.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000470116.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000489009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000489009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000489009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000580201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000580201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000580201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000607144.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000607144.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000607144.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | ENST00000607144.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002461.hg.1 | ENST00000225609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002461.hg.1 | ENST00000395844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002461.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002461.hg.1 | TC17000184.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700200876.hg.1 | TC1700012198.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000580201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | ENST00000580201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700200875.hg.1 | TC1700012198.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000225609.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000395844.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000463810.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000463810.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000463810.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000470116.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000470116.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000470116.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000470116.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000477745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000488375.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000489009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000489009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000489009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000498772.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000580201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000580201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000580201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000584797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000585034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000607144.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000607144.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000607144.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | ENST00000607144.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | NM_004278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012198.hg.1 | NM_004278.3 | Refseq |

PIGL
Genomic Map