Assay Details
Gene Symbol: PLGRKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018465.3 | NP_060935.2 | 2 - 3 | 276 | 93 | |
XM_005251510.4 | XP_005251567.1 | 2 - 3 | 391 | 93 | ||
XM_011517960.2 | XP_011516262.1 | 2 - 3 | 424 | 93 | ||
GenBank mRNA | AF220050.1 | - | 2 - 3 | 200 | 93 | |
AF225420.1 | - | 2 - 3 | 217 | 93 | ||
AK027594.1 | - | 2 - 3 | 215 | 93 | ||
AK312736.1 | - | 2 - 3 | 205 | 93 | ||
BC008212.1 | - | 2 - 3 | 186 | 3871917 | 93 | |
BM693747.1 | - | 2 - 3 | 276 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF220050.1, AF225420.1, AK027594.1, AK312736.1, BC008212.1, NM_018465.3, XM_005251510.4, XM_011517960.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
