Assay Details
| Gene Symbol: ABCC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF395908.1 | - | 3 - 4 | 433 | 69 | |
| AF395909.1 | - | 3 - 4 | 433 | 69 | ||
| AF411577.1 | - | 3 - 4 | 433 | 69 | ||
| AF411578.1 | - | 3 - 4 | 433 | 69 | ||
| AK122635.1 | - | 4 - 5 | 630 | 69 | ||
| AK302520.1 | - | 4 - 5 | 630 | 69 | ||
| AY196326.1 | - | 3 - 4 | 653 | 69 | ||
| BC036378.1 | - | 5 - 6 | 931 | 69 | ||
More Information
Set Membership: |
|

ABCC12
Genomic Map