Assay Details
| Gene Symbol: UNC80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005246476.1 | XP_005246533.1 | 19 - 20 | 3326 | 83 | |
| XM_011511006.1 | XP_011509308.1 | 19 - 20 | 3326 | 83 | ||
| XM_017003885.1 | XP_016859374.1 | 19 - 20 | 3326 | 83 | ||
| XM_017003888.1 | XP_016859377.1 | 19 - 20 | 3326 | 83 | ||
| XM_017003889.1 | XP_016859378.1 | 19 - 20 | 3326 | 83 | ||
| XM_017003890.1 | XP_016859379.1 | 19 - 20 | 3311 | 83 | ||
More Information
Set Membership: |
|

UNC80
Genomic Map