Assay Details
| Gene Symbol: EIF4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014740.3 | NP_055555.1 | 6 - 7 | 807 | 79 | |
| XM_011525522.1 | XP_011523824.1 | 5 - 6 | 700 | 79 | ||
| GenBank mRNA | AK290608.1 | - | 6 - 7 | 788 | 79 | |
| BC003662.2 | - | 6 - 7 | 717 | 2988931 | 79 | |
| BC004386.1 | - | 6 - 7 | 738 | 3619386 | 79 | |
| BC011151.1 | - | 6 - 7 | 742 | 3452439 | 79 | |
| CR456750.1 | - | 6 - 7 | 585 | 79 | ||
| D21853.1 | - | 6 - 7 | 799 | 79 | ||
| X79538.1 | - | 6 - 7 | 658 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001933.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
nuclear-transcribed mRNA poly(A) tail shortening
mRNA splicing, via spliceosome
rRNA processing
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
associative learning
RNA splicing
RNA secondary structure unwinding
response to organic cyclic compound
selenocysteine metabolic process
negative regulation of translation
mRNA 3'-end processing
exploration behavior
positive regulation of translation
embryonic cranial skeleton morphogenesis
cellular response to selenite ion
negative regulation of excitatory postsynaptic potential
negative regulation of selenocysteine incorporation
negative regulation of selenocysteine insertion sequence binding
cellular response to brain-derived neurotrophic factor stimulus
mRNA binding
ATP-dependent RNA helicase activity
protein binding
ATP binding
poly(A) binding
selenocysteine insertion sequence binding
RNA stem-loop binding
ribonucleoprotein complex binding
poly(A) RNA binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
nuclear-transcribed mRNA poly(A) tail shortening
mRNA splicing, via spliceosome
rRNA processing
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
associative learning
RNA splicing
RNA secondary structure unwinding
response to organic cyclic compound
selenocysteine metabolic process
negative regulation of translation
mRNA 3'-end processing
exploration behavior
positive regulation of translation
embryonic cranial skeleton morphogenesis
cellular response to selenite ion
negative regulation of excitatory postsynaptic potential
negative regulation of selenocysteine incorporation
negative regulation of selenocysteine insertion sequence binding
cellular response to brain-derived neurotrophic factor stimulus
mRNA binding
ATP-dependent RNA helicase activity
protein binding
ATP binding
poly(A) binding
selenocysteine insertion sequence binding
RNA stem-loop binding
ribonucleoprotein complex binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800008310.hg.1 | ENST00000578499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026342.hg.1 | ENST00000269349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026342.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026342.hg.1 | TC17001933.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000570625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000570625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000570625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000570625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000570837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000570837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000570837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000570837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000570625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000570625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000570625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000570625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000570837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000570837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000570837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000570837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | ENST00000576573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011967.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197739.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197739.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197739.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197739.hg.1 | ENST00000269349.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197739.hg.1 | ENST00000575978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197739.hg.1 | NM_014740.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197739.hg.1 | TC1700011967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1800008310.hg.1 | ENST00000578499.1 | Ensembl |

EIF4A3
Genomic Map