Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009677.hg.1 |
ENST00000344846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009677.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009677.hg.1 |
TC19000701.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000595322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000595322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000595322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000600863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000600863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000600863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
ENST00000595322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
ENST00000600863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
TC1900008469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
ENST00000595322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
ENST00000600863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165130.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000595322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000595322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000595322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000600863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000600863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000600863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
ENST00000601610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008469.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165126.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165126.hg.1 |
NM_012451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165126.hg.1 |
TC1900008469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165126.hg.1 |
ENST00000344846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165126.hg.1 |
NM_012451.3 |
Refseq |