Assay Details
| Gene Symbol: LRRC46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033413.3 | NP_219481.1 | 4 - 5 | 638 | 65 | |
| XM_005257775.4 | XP_005257832.1 | 3 - 4 | 351 | 65 | ||
| GenBank mRNA | AK027206.1 | - | 4 - 5 | 1441 | 65 | |
| AK292765.1 | - | 4 - 5 | 553 | 65 | ||
| AK315665.1 | - | 4 - 5 | 638 | 65 | ||
| BC010412.2 | - | 4 - 5 | 531 | 3836250 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000614.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007596.hg.1 | ENST00000269025.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007596.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007596.hg.1 | TC17000614.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700159939.hg.1 | TC1700008138.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |

LRRC46
Genomic Map