Assay Details
| Gene Symbol: ANKRD19P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026868.1 | 7 - 8 | 912 | 95 | ||
| GenBank mRNA | AK093497.1 | - | 5 - 6 | 504 | 95 | |
| AK292218.1 | - | 7 - 8 | 1325 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929748 | LOC101929748 | -17.9kb | Sense | Chr. 9 - 92805385 - 92808087 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 484 | ZNF484 | BA526D8.4 | 13.4kb | Sense | Chr. 9 - 92834262 - 92878060 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000438.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09004627.hg.1 | TC09000438.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004627.hg.1 | ENST00000473204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004627.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004628.hg.1 | TC09000438.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004628.hg.1 | ENST00000473204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004628.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004630.hg.1 | TC09000438.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004630.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184293.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184293.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184293.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184293.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184293.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184293.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184293.hg.1 | TC0900012151.hg.1 | Clariom D TC |

ANKRD19P
Genomic Map