| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004631.hg.1 |
TC09000438.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004631.hg.1 |
ENST00000473204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004631.hg.1 |
NR_026868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004634.hg.1 |
TC09000438.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004634.hg.1 |
ENST00000473204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004634.hg.1 |
NR_026868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004633.hg.1 |
TC09000438.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004633.hg.1 |
ENST00000473204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09004633.hg.1 |
NR_026868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184295.hg.1 |
ENST00000464387.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184295.hg.1 |
NR_026868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184295.hg.1 |
ENST00000464387.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184295.hg.1 |
NR_026868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184295.hg.1 |
TC0900012151.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900184296.hg.1 |
ENST00000464387.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184296.hg.1 |
NR_026868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184296.hg.1 |
ENST00000464387.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184296.hg.1 |
NR_026868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184296.hg.1 |
TC0900012151.hg.1 |
Clariom D TC |