Assay Details
| Gene Symbol: EEF1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004280.4 | NP_004271.1 | 3 - 4 | 455 | 174 | |
| GenBank mRNA | AB011079.1 | - | 3 - 4 | 381 | 174 | |
| AF054186.1 | - | 3 - 4 | 409 | 174 | ||
| AV714140.2 | - | 3 - 4 | 455 | 174 | ||
| BC005291.1 | - | 3 - 4 | 408 | 3685030 | 174 | |
| BT007306.1 | - | 3 - 4 | 381 | 174 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004111.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
tRNA aminoacylation for protein translation
glutathione metabolic process
negative regulation of cell proliferation
positive regulation of apoptotic process
positive regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of cellular senescence
translation elongation factor activity
glutathione transferase activity
protein binding
translational elongation
tRNA aminoacylation for protein translation
glutathione metabolic process
negative regulation of cell proliferation
positive regulation of apoptotic process
positive regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of cellular senescence
translation elongation factor activity
glutathione transferase activity
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000488226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000488226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000515633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000515633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000515633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | ENST00000515633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014969.hg.1 | TC06004111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014969.hg.1 | ENST00000379715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014969.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008893.hg.1 | ENST00000446998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008893.hg.1 | ENST00000446998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008893.hg.1 | ENST00000446998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000429723.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000488226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000488226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000502429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000507463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000515633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000515633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000515633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | ENST00000515633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_001135650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014239.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200692.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200692.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200692.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200692.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200692.hg.1 | TC0600014239.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008893.hg.1 | ENST00000446998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008893.hg.1 | ENST00000446998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008893.hg.1 | ENST00000446998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200693.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200693.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200693.hg.1 | ENST00000379715.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200693.hg.1 | NM_004280.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200693.hg.1 | TC0600014239.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 191646 | - | - |

EEF1E1
Genomic Map