Assay Details
| Gene Symbol: MYO5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK025336.1 | - | 9 - 10 | 1376 | 96 | |
| AL133643.1 | - | 8 - 9 | 1010 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYO5B| Gene Symbol: MYO5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK025336.1 | - | 9 - 10 | 1376 | 96 | |
| AL133643.1 | - | 8 - 9 | 1010 | 96 | ||
Set Membership: |
|