Assay Details
| Gene Symbol: MBOAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080480.2 | NP_001073949.1 | 1 - 2 | 330 | 112 | |
| XM_006715000.3 | XP_006715063.1 | 1 - 2 | 268 | 112 | ||
| XM_011514313.2 | XP_011512615.1 | 1 - 2 | 268 | 112 | ||
| GenBank mRNA | AB305043.1 | - | 1 - 2 | 100 | 112 | |
| AK131269.1 | - | 1 - 2 | 293 | 112 | ||
| AK304748.1 | - | 1 - 2 | 275 | 112 | ||
| BC150650.1 | - | 1 - 2 | 276 | 9057021 | 112 | |
| BC150652.1 | - | 1 - 2 | 276 | 9057023 | 112 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001313.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06015735.hg.1 | TC06001313.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015735.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015735.hg.1 | ENST00000536798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015735.hg.1 | ENST00000541730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015735.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015734.hg.1 | TC06001313.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015734.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015734.hg.1 | ENST00000536798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015734.hg.1 | ENST00000541730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015734.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015737.hg.1 | TC06001313.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015737.hg.1 | ENST00000536798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015737.hg.1 | ENST00000541730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015737.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015733.hg.1 | TC06001313.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015733.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015733.hg.1 | ENST00000536798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015733.hg.1 | ENST00000541730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015733.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174557.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174557.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174557.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174557.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174557.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174557.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174557.hg.1 | TC0600010991.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600174556.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174556.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174556.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174556.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174556.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174556.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174556.hg.1 | TC0600010991.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010991.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174555.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174555.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174555.hg.1 | ENST00000324607.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174555.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174555.hg.1 | TC0600010991.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600174559.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174559.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174559.hg.1 | NM_001080480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174559.hg.1 | NR_073465.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174559.hg.1 | TC0600010991.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 182575 | - | - |

MBOAT1
Genomic Map