Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000253496.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000502854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000503736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000503736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000503736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000503736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000503736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000503736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000504406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000504406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000504406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000504406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000504406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000504406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000504406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000504406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000504406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000510358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000514943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000514943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000514943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
ENST00000514943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012971.hg.1 |
NM_000505.3 |
Refseq |