Assay Details
| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000505.3 | NP_000496.2 | 4 - 5 | 341 | 129 | |
| GenBank mRNA | AB095845.1 | - | 4 - 5 | 341 | 129 | |
| M11723.1 | - | 4 - 5 | 253 | 129 | ||
| M31315.1 | - | 4 - 5 | 283 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05002105.hg.1
Set Membership: |
|

F12
Genomic Map