Assay Details
| Gene Symbol: CLCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256944.1 | NP_001243873.1 | 4 - 5 | 728 | 74 | |
| NM_001830.3 | NP_001821.2 | 6 - 7 | 983 | 74 | ||
| GenBank mRNA | AB019432.1 | - | 5 - 6 | 828 | 74 | |
| AF170492.1 | - | 4 - 5 | 553 | 74 | ||
| AK225287.1 | - | 6 - 7 | 944 | 74 | ||
| AK289564.1 | - | 6 - 7 | 930 | 74 | ||
| AK299611.1 | - | 3 - 4 | 361 | 74 | ||
| BC036068.1 | - | 5 - 6 | 603 | 5311366 | 74 | |
| BC130278.1 | - | 5 - 6 | 842 | 40146309 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000038.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000527.hg.1 | TC0X000038.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000527.hg.1 | ENST00000380829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000527.hg.1 | ENST00000380833.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000527.hg.1 | ENST00000421085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000527.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145070.hg.1 | TC0X00006593.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |

CLCN4
Genomic Map