| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |