Assay Details
| Gene Symbol: N4BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534473.1 | XP_011532775.1 | 5 - 6 | 1972 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

N4BP3| Gene Symbol: N4BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534473.1 | XP_011532775.1 | 5 - 6 | 1972 | 103 | |
Set Membership: |
|