Assay Details
| Gene Symbol: LOC105373132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544354.2 | XP_011542656.1 | 1 | 3434 | 145 | |
| Gene Symbol: DUSP5P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055963.1 | - | 1 | 2104 | 145 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RNA, 5S ribosomal 17 |
RNA5S17 | RN5S17 | -1.5kb | Anti-sense | Chr. 1 - 228646038 - 228646158 on NCBI Build GRCh38 |
| Downstream |
RAB4A, member RAS oncogene family |
RAB4A |
HRES-1, HRES-1/RAB4, HRES1, RAB4 |
621.1kb | Sense | Chr. 1 - 229271062 - 229305894 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105373132
Genomic Map