Assay Details
| Gene Symbol: ADCY9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001116.3 | NP_001107.2 | 6 - 7 | 2840 | 79 | |
| XM_005255079.3 | XP_005255136.1 | 6 - 7 | 2722 | 79 | ||
| XM_011522353.2 | XP_011520655.1 | 6 - 7 | 2724 | 79 | ||
| GenBank mRNA | AB011092.3 | - | 6 - 7 | 2681 | 79 | |
| AF036927.1 | - | 5 - 6 | 2317 | 79 | ||
| AJ133123.1 | - | 6 - 7 | 2839 | 79 | ||
| BC136657.1 | - | 5 - 6 | 2357 | 9020647 | 79 | |
| BC136658.1 | - | 5 - 6 | 2357 | 9020648 | 79 | |
| BC151207.1 | - | 6 - 7 | 2676 | 40147127 | 79 | |
| BC151229.1 | - | 6 - 7 | 2681 | 9007273 | 79 | |
| DQ005545.1 | - | 5 - 6 | 2301 | 79 | ||
| DQ008441.1 | - | 5 - 6 | 2301 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000824.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
renal water homeostasis
cAMP biosynthetic process
signal transduction
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
cAMP-mediated signaling
activation of protein kinase A activity
cellular response to glucagon stimulus
adenylate cyclase-activating adrenergic receptor signaling pathway
adenylate cyclase activity
ATP binding
metal ion binding
renal water homeostasis
cAMP biosynthetic process
signal transduction
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
cAMP-mediated signaling
activation of protein kinase A activity
cellular response to glucagon stimulus
adenylate cyclase-activating adrenergic receptor signaling pathway
adenylate cyclase activity
ATP binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16011957.hg.1 | ENST00000294016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011957.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011957.hg.1 | TC16000824.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170415.hg.1 | TC1600009210.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170414.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170414.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170414.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170414.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170414.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170414.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170414.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170414.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170414.hg.1 | TC1600009210.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |

ADCY9
Genomic Map