Assay Details
| Gene Symbol: BTNL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040462.2 | NP_001035552.1 | 4 - 5 | 1026 | 77 | |
| NM_001159707.1 | NP_001153179.1 | 3 - 4 | 678 | 77 | ||
| NM_001159709.1 | NP_001153181.1 | 4 - 5 | 729 | 77 | ||
| NM_001159710.1 | NP_001153182.1 | 3 - 4 | 529 | 77 | ||
| XM_011534650.2 | XP_011532952.1 | 4 - 5 | 831 | 77 | ||
| XM_011534651.2 | XP_011532953.1 | 3 - 4 | 1845 | 77 | ||
| XM_011534652.2 | XP_011532954.1 | 3 - 4 | 1821 | 77 | ||
| XM_011534653.2 | XP_011532955.1 | 2 - 3 | 382 | 77 | ||
| GenBank mRNA | AK296597.1 | - | 4 - 5 | 684 | 77 | |
| AK298687.1 | - | 3 - 4 | 529 | 77 | ||
| AY358523.1 | - | 4 - 5 | 974 | 77 | ||
| BC119696.1 | - | 3 - 4 | 481 | 40116820 | 77 | |
More Information
Set Membership: |
|

BTNL8
Genomic Map