Assay Details
| Gene Symbol: BTNL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040462.2 | NP_001035552.1 | 5 - 6 | 1047 | 105 | |
| NM_001159707.1 | NP_001153179.1 | 4 - 5 | 699 | 105 | ||
| NM_001159709.1 | NP_001153181.1 | 5 - 6 | 750 | 105 | ||
| NM_001159710.1 | NP_001153182.1 | 4 - 5 | 550 | 105 | ||
| NM_024850.2 | NP_079126.1 | 5 - 6 | 1177 | 235 | ||
| XM_011534650.2 | XP_011532952.1 | 5 - 6 | 852 | 105 | ||
| XM_011534651.2 | XP_011532953.1 | 4 - 5 | 1866 | 105 | ||
| XM_011534652.2 | XP_011532954.1 | 4 - 5 | 1842 | 105 | ||
| XM_011534653.2 | XP_011532955.1 | 3 - 4 | 403 | 105 | ||
| GenBank mRNA | AK025111.1 | - | 5 - 6 | 1095 | 235 | |
| AK225213.1 | - | 5 - 6 | 1095 | 235 | ||
| AK296597.1 | - | 5 - 6 | 705 | 105 | ||
| AK298687.1 | - | 4 - 5 | 550 | 105 | ||
| AY358523.1 | - | 5 - 6 | 995 | 105 | ||
| BC119696.1 | - | 4 - 5 | 502 | 40116820 | 105 | |
More Information
Set Membership: |
|

BTNL8
Genomic Map