Assay Details
| Gene Symbol: ANKRD19P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC028712.1 | - | 7 - 8 | 1049 | 145 | |
| BC038951.1 | - | 6 - 7 | 859 | 5733799 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184291.hg.1 | TC0900012151.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012151.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000338192.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000460854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000464387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000465736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | ENST00000492083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | NR_026868.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184290.hg.1 | TC0900012151.hg.1 | Clariom D TC |

ANKRD19P
Genomic Map