Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027092.hg.1 |
TC01001733.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027092.hg.1 |
ENST00000367070.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027092.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183887.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183887.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183887.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183887.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183887.hg.1 |
TC0100011402.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000367070.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000421786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
ENST00000424088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011402.hg.1 |
NM_000715.3 |
Refseq |