Assay Details
| Gene Symbol: C4BPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000715.3 | NP_000706.1 | 1 - 2 | 171 | 89 | |
| GenBank mRNA | AK313164.1 | - | 1 - 2 | 117 | 89 | |
| BC022312.1 | - | 1 - 2 | 171 | 4688428 | 89 | |
| M31452.1 | - | 1 - 2 | 115 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001733.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK313164.1,BC022312.1,M31452.1,NM_000715.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01027079.hg.1 | TC01001733.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027079.hg.1 | ENST00000367070.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027079.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027080.hg.1 | TC01001733.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027080.hg.1 | ENST00000367070.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027080.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027082.hg.1 | TC01001733.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027082.hg.1 | ENST00000367070.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027082.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027078.hg.1 | TC01001733.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027078.hg.1 | ENST00000367070.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027078.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183870.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183870.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183870.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183870.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183870.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183870.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183870.hg.1 | TC0100011402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100183875.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183875.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183875.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183875.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183875.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183875.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183875.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183875.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183875.hg.1 | TC0100011402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100183869.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183869.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183869.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183869.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183869.hg.1 | TC0100011402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100183871.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183871.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183871.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183871.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183871.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183871.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183871.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183871.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183871.hg.1 | TC0100011402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000367070.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000421786.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | ENST00000424088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011402.hg.1 | NM_000715.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 213801 | - | - |

C4BPA
Genomic Map