Assay Details
| Gene Symbol: ZSWIM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020928.1 | NP_065979.1 | 2 - 3 | 1034 | 118 | |
| GenBank mRNA | BF750632.1 | - | 1 - 2 | 253 | 118 | |
| BF751445.1 | - | 1 - 2 | 261 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000262.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05003175.hg.1 | TC05000262.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003175.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003175.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003173.hg.1 | TC05000262.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003173.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003173.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003174.hg.1 | TC05000262.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003174.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003174.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500150846.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500150846.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500150846.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500150846.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500150846.hg.1 | TC0500007549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500150850.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500150850.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500150850.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500150850.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500150850.hg.1 | TC0500007549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500150848.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500150848.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500150848.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500150848.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500150848.hg.1 | TC0500007549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | ENST00000252744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007549.hg.1 | NM_020928.1 | Refseq |

ZSWIM6
Genomic Map