Assay Details
| Gene Symbol: COL15A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001855.4 | NP_001846.3 | 39 - 40 | 3996 | 77 | |
| XM_011518214.2 | XP_011516516.1 | 40 - 41 | 3991 | 77 | ||
| GenBank mRNA | L01697.1 | - | 29 - 30 | 2018 | 77 | |
| L25286.1 | - | 39 - 40 | 3736 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000499.hg.1
Set Membership: |
|

COL15A1
Genomic Map