Assay Details
| Gene Symbol: SIRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193286.1 | NP_001180215.1 | 2 - 3 | 407 | 94 | |
| NM_030593.2 | NP_085096.1 | 2 - 3 | 407 | 94 | ||
| XM_017026500.1 | XP_016881989.1 | 2 - 3 | 171 | 94 | ||
| GenBank mRNA | AF095714.1 | - | 2 - 3 | 186 | 94 | |
| AF131800.1 | - | 1 - 2 | 62 | 94 | ||
| AK025876.1 | - | 2 - 3 | 193 | 94 | ||
| AK225824.1 | - | 2 - 3 | 190 | 94 | ||
| AY030277.1 | - | 2 - 3 | 168 | 94 | ||
| BC003012.1 | - | 2 - 3 | 258 | 2820929 | 94 | |
| BC003547.1 | - | 2 - 3 | 258 | 3536728 | 94 | |
| DA541045.1 | - | 2 - 3 | 408 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF095714.1, AY030277.1, BC003012.1, BC003547.1, NM_001193286.1, NM_030593.2, XM_017026500.1 transcripts.
Set Membership: |
|

SIRT2
Genomic Map