Assay Details
Gene Symbol: SYNGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012451.3 | NP_036583.2 | 1 - 2 | 148 | 153 | |
GenBank mRNA | BG720827.1 | - | 1 - 2 | 150 | 153 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000701.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_012451.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19009675.hg.1 | ENST00000344846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009675.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009675.hg.1 | TC19000701.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19009673.hg.1 | ENST00000344846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009673.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009673.hg.1 | TC19000701.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000595322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000595322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000595322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000600863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000600863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000600863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009676.hg.1 | ENST00000344846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009676.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009676.hg.1 | TC19000701.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900165124.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900165124.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900165124.hg.1 | TC1900008469.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900165124.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900165124.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000595322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000595322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000595322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000600863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000600863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000600863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | ENST00000601610.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008469.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900165123.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900165123.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900165123.hg.1 | TC1900008469.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900165123.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900165123.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900165125.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900165125.hg.1 | NM_012451.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900165125.hg.1 | TC1900008469.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900165125.hg.1 | ENST00000344846.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900165125.hg.1 | NM_012451.3 | Refseq |
