Assay Details
| Gene Symbol: DNAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK074130.2 | - | 23 - 24 | 4234 | 64 | |
| AY221994.1 | - | 32 - 33 | 5752 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAH1| Gene Symbol: DNAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK074130.2 | - | 23 - 24 | 4234 | 64 | |
| AY221994.1 | - | 32 - 33 | 5752 | 64 | ||
Set Membership: |
|