Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021415.hg.1 |
TC06001776.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021415.hg.1 |
ENST00000283297.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021415.hg.1 |
ENST00000371253.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021415.hg.1 |
ENST00000449332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06021415.hg.1 |
NM_153840.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
ENST00000283297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
ENST00000371253.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
ENST00000419892.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
ENST00000449332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
NM_153840.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
ENST00000283297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
ENST00000371253.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
ENST00000419892.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
ENST00000449332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
NM_153840.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182353.hg.1 |
TC0600011957.hg.1 |
Clariom D TC |