Assay Details
| Gene Symbol: RAB23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278666.1 | NP_001265595.1 | 4 - 5 | 590 | 112 | |
| NM_001278667.1 | NP_001265596.1 | 4 - 5 | 762 | 112 | ||
| NM_001278668.1 | NP_001265597.1 | 4 - 5 | 589 | 112 | ||
| NM_016277.4 | NP_057361.3 | 4 - 5 | 1059 | 112 | ||
| NM_183227.2 | NP_899050.1 | 4 - 5 | 613 | 112 | ||
| GenBank mRNA | AB025427.1 | - | 3 - 4 | 405 | 112 | |
| AB034244.1 | - | 4 - 5 | 558 | 112 | ||
| AF498951.1 | - | 3 - 4 | 405 | 112 | ||
| AK313796.1 | - | 4 - 5 | 610 | 112 | ||
| AY585189.1 | - | 3 - 4 | 405 | 112 | ||
| BC015021.1 | - | 4 - 5 | 1025 | 3910708 | 112 | |
| CR749371.1 | - | 5 - 6 | 793 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001830.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022308.hg.1 | TC06001830.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022308.hg.1 | ENST00000317483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022308.hg.1 | ENST00000344445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022308.hg.1 | ENST00000468148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022308.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022308.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600183738.hg.1 | TC0600012132.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000317483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | ENST00000468148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278666.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278667.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_001278668.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_016277.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NM_183227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NR_103822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012132.hg.1 | NR_103822.1 | Refseq |

RAB23
Genomic Map