Assay Details
| Gene Symbol: ABCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004996.3 | NP_004987.2 | 19 - 20 | 2823 | 79 | |
| XM_011522497.1 | XP_011520799.1 | 19 - 20 | 2639 | 79 | ||
| XM_011522498.2 | XP_011520800.1 | 18 - 19 | 2558 | 79 | ||
| XM_017023237.1 | XP_016878726.1 | 19 - 20 | 2705 | 79 | ||
| XM_017023238.1 | XP_016878727.1 | 18 - 19 | 2579 | 79 | ||
| XM_017023239.1 | XP_016878728.1 | 18 - 19 | 2567 | 79 | ||
| XM_017023240.1 | XP_016878729.1 | 18 - 19 | 2528 | 79 | ||
| XM_017023241.1 | XP_016878730.1 | 17 - 18 | 2441 | 79 | ||
| XM_017023242.1 | XP_016878731.1 | 17 - 18 | 2360 | 79 | ||
| XM_017023243.1 | XP_016878732.1 | 19 - 20 | 2705 | 79 | ||
| GenBank mRNA | L05628.1 | - | 19 - 20 | 2844 | 79 | |
| X78338.1 | - | 19 - 20 | 2785 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
leukotriene metabolic process
transport
cobalamin metabolic process
cobalamin transport
vitamin transmembrane transport
response to drug
transmembrane transport
anion transmembrane transport
transporter activity
ATP binding
cobalamin-transporting ATPase activity
ATPase activity
ATPase activity, coupled to transmembrane movement of substances
anion transmembrane-transporting ATPase activity
leukotriene metabolic process
transport
cobalamin metabolic process
cobalamin transport
vitamin transmembrane transport
response to drug
transmembrane transport
anion transmembrane transport
transporter activity
ATP binding
cobalamin-transporting ATPase activity
ATPase activity
ATPase activity, coupled to transmembrane movement of substances
anion transmembrane-transporting ATPase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000568853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000568853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | ENST00000568853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000499217.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000564621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000567286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000568853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000568853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | ENST00000568853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038448.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_038449.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007848.hg.1 | NR_120377.1 | Refseq |

ABCC1
Genomic Map