Assay Details
| Gene Symbol: ABCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB209120.1 | - | 18 - 19 | 2373 | 83 | |
| CR749835.1 | - | 16 - 17 | 2131 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCC1| Gene Symbol: ABCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB209120.1 | - | 18 - 19 | 2373 | 83 | |
| CR749835.1 | - | 16 - 17 | 2131 | 83 | ||
Set Membership: |
|