Assay Details
| Gene Symbol: DDX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006773.3 | NP_006764.3 | 13 - 14 | 1973 | 90 | |
| GenBank mRNA | AB209392.1 | - | 4 - 5 | 3889 | 90 | |
| AK001467.1 | - | 13 - 14 | 1945 | 90 | ||
| AK091227.1 | - | 11 - 12 | 1400 | 90 | ||
| BC001238.1 | - | 13 - 14 | 1902 | 3452935 | 90 | |
| BC003360.1 | - | 13 - 14 | 1932 | 2900252 | 90 | |
| BC024739.1 | - | 13 - 14 | 1947 | 4995871 | 90 | |
| CR457060.1 | - | 12 - 13 | 1694 | 90 | ||
| X98743.1 | - | 12 - 13 | 1711 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000742.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02011585.hg.1 | TC02000742.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011585.hg.1 | ENST00000263239.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011585.hg.1 | ENST00000539346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011585.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011584.hg.1 | TC02000742.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011584.hg.1 | ENST00000263239.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011584.hg.1 | ENST00000539346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011584.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200163235.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163235.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163235.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200163235.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163235.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163235.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200163235.hg.1 | TC0200009041.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200163236.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163236.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163236.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200163236.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163236.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163236.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200163236.hg.1 | TC0200009041.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200163234.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163234.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163234.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200163234.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163234.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200163234.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200163234.hg.1 | TC0200009041.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000263239.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000415038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000461443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000474694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000476149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | ENST00000489933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009041.hg.1 | NM_006773.3 | Refseq |

DDX18
Genomic Map