Assay Details
| Gene Symbol: APP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136129.2 | NP_001129601.1 | 5 - 6 | 897 | 132 | |
| NM_001136131.2 | NP_001129603.1 | 6 - 7 | 952 | 132 | ||
| NM_001204303.1 | NP_001191232.1 | 6 - 7 | 1065 | 132 | ||
| NM_201414.2 | NP_958817.1 | 6 - 7 | 1065 | 132 | ||
| GenBank mRNA | A02759.1 | - | 6 - 7 | 1011 | 132 | |
| AF282245.1 | - | 5 - 6 | 824 | 132 | ||
| AK296229.1 | - | 6 - 7 | 952 | 132 | ||
| AK297412.1 | - | 5 - 6 | 794 | 132 | ||
| AK298861.1 | - | 5 - 6 | 967 | 132 | ||
| Y00264.1 | - | 6 - 7 | 1012 | 132 | ||
More Information
Set Membership: |
|

APP
Genomic Map