| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146107.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146107.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146107.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146107.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146107.hg.1 |
TC1900006603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900146107.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146107.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146107.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146107.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
TC1900006603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146105.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146106.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146106.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146106.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146106.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146106.hg.1 |
TC1900006603.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900146106.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146106.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146106.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146106.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000307635.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586426.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
ENST00000586470.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_001300843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006603.hg.1 |
NM_024967.2 |
Refseq |