Assay Details
| Gene Symbol: KCNAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172130.2 | NP_742128.1 | 2 - 3 | 561 | 108 | |
| XM_005263514.2 | XP_005263571.1 | 2 - 3 | 316 | 108 | ||
| GenBank mRNA | AK312871.1 | - | 2 - 3 | 241 | 108 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000084.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312871.1,NM_172130.2,XM_005263514.2 transcripts.
Set Membership: |
|

KCNAB2
Genomic Map