Assay Details
| Gene Symbol: KRT32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002278.3 | NP_002269.3 | 4 - 5 | 928 | 57 | |
| GenBank mRNA | X81419.1 | - | 4 - 5 | 767 | 57 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001505.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17019835.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019835.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019835.hg.1 | TC17001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019836.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019836.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019836.hg.1 | TC17001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019833.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019833.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019833.hg.1 | TC17001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019831.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019831.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019831.hg.1 | TC17001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019834.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019834.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019834.hg.1 | TC17001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019832.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019832.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019832.hg.1 | TC17001505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184335.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184335.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184335.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184335.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184335.hg.1 | TC1700010672.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184340.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184340.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184340.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184340.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184340.hg.1 | TC1700010672.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184336.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184336.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184336.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184336.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184336.hg.1 | TC1700010672.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010672.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184334.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184334.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184334.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184334.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184334.hg.1 | TC1700010672.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184338.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184338.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184338.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184338.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184338.hg.1 | TC1700010672.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184337.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184337.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184337.hg.1 | ENST00000225899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184337.hg.1 | NM_002278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184337.hg.1 | TC1700010672.hg.1 | Clariom D TC |

KRT32
Genomic Map